Product Details

SNP ID
rs200078797
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28325579 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGAGCCCCCAGCCTGGTTTAAGT[C/T]CTTTCTGTTTTGCGAGCTTGTGTTT
Phenotype
MIM: 614394 MIM: 612912 MIM: 191161
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IFT20 PubMed Links

Gene Details

Gene
IFT20
Gene Name
intraflagellar transport 20
There are no transcripts associated with this gene.

Gene
TMEM97
Gene Name
transmembrane protein 97
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014573.2 397 Missense Mutation TCC,TTC S68F NP_055388.2
XM_005257965.3 397 Missense Mutation TCC,TTC S107F XP_005258022.1
Gene
TNFAIP1
Gene Name
TNF alpha induced protein 1
There are no transcripts associated with this gene.

View Full Product Details