Product Details

SNP ID
rs200384262
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:45999662 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAGTCAGGCGACCTCATGCCAAG[C/T]GTAGAAAGGGGCAGATGGGAGCCCC
Phenotype
MIM: 157140 MIM: 607067
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAPT PubMed Links

Gene Details

Gene
MAPT
Gene Name
microtubule associated protein tau
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001123066.3 413 Intron NP_001116538.2
NM_001123067.3 413 Intron NP_001116539.1
NM_001203251.1 413 Intron NP_001190180.1
NM_001203252.1 413 Intron NP_001190181.1
NM_005910.5 413 Intron NP_005901.2
NM_016834.4 413 Intron NP_058518.1
NM_016835.4 413 Intron NP_058519.3
NM_016841.4 413 Intron NP_058525.1
XM_005257362.4 413 Intron XP_005257419.1
XM_005257364.4 413 Intron XP_005257421.1
XM_005257365.4 413 Intron XP_005257422.1
XM_005257366.3 413 Intron XP_005257423.1
XM_005257367.4 413 Intron XP_005257424.1
XM_005257368.4 413 Intron XP_005257425.1
XM_005257369.4 413 Intron XP_005257426.1
XM_005257370.4 413 Intron XP_005257427.1
XM_005257371.4 413 Intron XP_005257428.1
Gene
STH
Gene Name
saitohin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007532.2 413 Missense Mutation GCG,GTG A128V NP_001007533.1

View Full Product Details