Product Details

SNP ID
rs200576429
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:55720776 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCACAAGGCACTGGAGGCTAGTA[A/G]AAAAAGTCCAGATGACAGAACAACC
Phenotype
MIM: 616334
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM100 PubMed Links

Gene Details

Gene
TMEM100
Gene Name
transmembrane protein 100
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099640.1 2461 Silent Mutation CTA,TTA L99L NP_001093110.1
NM_018286.2 2461 Silent Mutation CTA,TTA L99L NP_060756.2
XM_017024814.1 2461 Silent Mutation CTA,TTA L99L XP_016880303.1
XM_017024815.1 2461 Silent Mutation CTA,TTA L99L XP_016880304.1
XM_017024816.1 2461 Silent Mutation CTA,TTA L99L XP_016880305.1

View Full Product Details