Product Details

SNP ID
rs199737978
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41930802 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAAGGCGAGCGGCTCTACCTGTGC[A/G]GGGAATTTTCTAAAGCCGCGCAGAG
Phenotype
MIM: 108728
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACLY PubMed Links

Gene Details

Gene
ACLY
Gene Name
ATP citrate lyase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001096.2 180 Intron NP_001087.2
NM_001303274.1 180 Intron NP_001290203.1
NM_001303275.1 180 Intron NP_001290204.1
NM_198830.1 180 Intron NP_942127.1
XM_005257395.1 180 Intron XP_005257452.1
XM_017024688.1 180 Intron XP_016880177.1
Gene
TTC25
Gene Name
tetratricopeptide repeat domain 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031421.3 180 Missense Mutation AGG,GGG R27G NP_113609.1
XM_011525323.2 180 UTR 5 XP_011523625.1
XM_011525324.2 180 Missense Mutation AGG,GGG R27G XP_011523626.1

View Full Product Details