Product Details
- SNP ID
-
rs200114460
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:28703850 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAAGGCTCCAATTTAACCGGGCTT[C/T]TCTTCTTAGTCAACTGGCCTTTCTT
- Phenotype
-
MIM: 601333
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PROCA1
PubMed Links
Gene Details
- Gene
- PROCA1
- Gene Name
- protein interacting with cyclin A1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001304949.1 |
933 |
Missense Mutation |
AAA,AGA |
K231R |
NP_001291878.1 |
NM_001304951.1 |
933 |
Missense Mutation |
AAA,AGA |
K124R |
NP_001291880.1 |
NM_001304952.1 |
933 |
Missense Mutation |
AAA,AGA |
K242R |
NP_001291881.1 |
NM_001304953.1 |
933 |
Missense Mutation |
AAA,AGA |
K168R |
NP_001291882.1 |
NM_001304954.1 |
933 |
Missense Mutation |
AAA,AGA |
K168R |
NP_001291883.1 |
NM_152465.2 |
933 |
Missense Mutation |
AAA,AGA |
K240R |
NP_689678.1 |
XM_005257927.4 |
933 |
Missense Mutation |
AAA,AGA |
K268R |
XP_005257984.1 |
XM_006721723.3 |
933 |
Missense Mutation |
AAA,AGA |
K265R |
XP_006721786.1 |
XM_011524398.2 |
933 |
Missense Mutation |
AAA,AGA |
K259R |
XP_011522700.1 |
XM_011524399.2 |
933 |
Missense Mutation |
AAA,AGA |
K231R |
XP_011522701.1 |
XM_011524400.2 |
933 |
Missense Mutation |
AAA,AGA |
K214R |
XP_011522702.1 |
XM_011524401.1 |
933 |
Missense Mutation |
AAA,AGA |
K168R |
XP_011522703.1 |
XM_017024237.1 |
933 |
Missense Mutation |
AAA,AGA |
K168R |
XP_016879726.1 |
- Gene
- SUPT6H
- Gene Name
- SPT6 homolog, histone chaperone
There are no transcripts associated with this gene.
View Full Product Details