Product Details

SNP ID
rs199503836
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:83085185 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGACGGGGACGGCAGGCCCGGC[C/T]GGGTGCAGTGTTTTGGCCTGGAGCA
Phenotype
MIM: 616241
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
METRNL PubMed Links

Gene Details

Gene
METRNL
Gene Name
meteorin like, glial cell differentiation regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004431.2 892 Silent Mutation CGG,TGG R140W NP_001004431.1
XM_017024523.1 892 Silent Mutation CGG,TGG R58W XP_016880012.1

View Full Product Details