Product Details

SNP ID
rs199911286
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:36013820 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGAACACGGGATGCTTCGTGGGG[C/T]GTAGGAGATGCAGCAGTCAGCACTA
Phenotype
MIM: 602494
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCL23 PubMed Links

Gene Details

Gene
CCL23
Gene Name
C-C motif chemokine ligand 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005064.5 297 Missense Mutation ACC,GCC T76A NP_005055.3
NM_145898.3 297 Missense Mutation ACC,GCC T59A NP_665905.2

View Full Product Details