Product Details

SNP ID
rs200048655
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:29571968 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGAGGAAGCAGAGGAAGAAGAAG[G/T]CATGGATCTACTGTGAAAGCCTTTC
Phenotype
MIM: 608434
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ABHD15 PubMed Links

Gene Details

Gene
ABHD15
Gene Name
abhydrolase domain containing 15
There are no transcripts associated with this gene.

Gene
GIT1
Gene Name
GIT ArfGAP 1
There are no transcripts associated with this gene.

Gene
TP53I13
Gene Name
tumor protein p53 inducible protein 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138349.2 462 Missense Mutation GCA,TCA A142S NP_612358.3
XM_017025282.1 462 Missense Mutation GCA,TCA A117S XP_016880771.1

View Full Product Details