Product Details

SNP ID
rs199673549
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42844734 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTGGTTCCAGCCAGCCTCCCCA[C/T]TGCCCCTCTGTATCCCATAGGGCCC
Phenotype
MIM: 602268 MIM: 603735 MIM: 605129
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AOC2 PubMed Links

Gene Details

Gene
AOC2
Gene Name
amine oxidase, copper containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001158.3 143 Silent Mutation CAC,CAT H36H NP_001149.2
NM_009590.2 143 Silent Mutation CAC,CAT H36H NP_033720.2
Gene
AOC3
Gene Name
amine oxidase, copper containing 3
There are no transcripts associated with this gene.

Gene
PSME3
Gene Name
proteasome activator subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267045.1 143 Intron NP_001253974.1
NM_005789.3 143 Intron NP_005780.2
NM_176863.2 143 Intron NP_789839.1
XM_011524177.2 143 Intron XP_011522479.1

View Full Product Details