Product Details

SNP ID
rs199573317
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:5214965 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGCCAATTCCTCGGTGGCAGAGG[C/T]GGTAATTGAACTGGCGACTCATTAA
Phenotype
MIM: 614406
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100130950 PubMed Links

Gene Details

Gene
LOC100130950
Gene Name
uncharacterized LOC100130950
There are no transcripts associated with this gene.

Gene
SCIMP
Gene Name
SLP adaptor and CSK interacting membrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271842.1 348 Silent Mutation CCA,CCG P74P NP_001258771.1
NM_001319190.1 348 Silent Mutation CCA,CCG P74P NP_001306119.1
NM_207103.3 348 Silent Mutation CCA,CCG P81P NP_996986.1
XM_017024615.1 348 Silent Mutation CCA,CCG P81P XP_016880104.1

View Full Product Details