Product Details

SNP ID
rs199745454
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:27880306 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGATCCAGTCAGCATCTTCAATGG[C/T]TCTTTTAATAATCTGCTGGATTCTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LYRM9 PubMed Links

Gene Details

Gene
LYRM9
Gene Name
LYR motif containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001076680.1 272 Missense Mutation ACC,GCC T63A NP_001070148.1
XM_011524491.2 272 Missense Mutation ACC,GCC T89A XP_011522793.1
XM_017024325.1 272 Missense Mutation ACC,GCC T63A XP_016879814.1

View Full Product Details