Product Details

SNP ID
rs199917421
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41988885 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCATCTTCTTTCTTTGCTTTGAG[G/T]GCTTTGGCATTCTACAGAAAAAAGC
Phenotype
MIM: 601964
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC7 PubMed Links

Gene Details

Gene
DNAJC7
Gene Name
DnaJ heat shock protein family (Hsp40) member C7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144766.2 936 Silent Mutation GCA,GCC A199A NP_001138238.1
NM_003315.3 936 Silent Mutation GCA,GCC A255A NP_003306.3
XM_011525167.2 936 Silent Mutation GCA,GCC A199A XP_011523469.1
XM_011525168.2 936 Silent Mutation GCA,GCC A199A XP_011523470.1
XM_011525169.2 936 Intron XP_011523471.1
XM_017024994.1 936 Silent Mutation GCA,GCC A199A XP_016880483.1

View Full Product Details