Product Details

SNP ID
rs200023716
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7592834 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTTCCCCAGGTTCTGAATCAACC[A/G]GGGGCTCTGGTTCAGACGTGTCCAA
Phenotype
MIM: 605339 MIM: 604041 MIM: 601297
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FXR2 PubMed Links

Gene Details

Gene
FXR2
Gene Name
FMR1 autosomal homolog 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004860.3 1954 Missense Mutation CCG,CTG P530L NP_004851.2
Gene
LOC100996842
Gene Name
uncharacterized LOC100996842
There are no transcripts associated with this gene.

Gene
MPDU1
Gene Name
mannose-P-dolichol utilization defect 1
There are no transcripts associated with this gene.

Gene
SOX15
Gene Name
SRY-box 15
There are no transcripts associated with this gene.

View Full Product Details