Product Details

SNP ID
rs199513433
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42114583 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTTGCGGATCTGGGCCTGTTTGC[A/G]CTCAATCAGCTTCTTGATGATCTGA
Phenotype
MIM: 608588 MIM: 608344 MIM: 602301
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DHX58 PubMed Links

Gene Details

Gene
DHX58
Gene Name
DEXH-box helicase 58
There are no transcripts associated with this gene.

Gene
HSPB9
Gene Name
heat shock protein family B (small) member 9
There are no transcripts associated with this gene.

Gene
KAT2A
Gene Name
lysine acetyltransferase 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021078.2 2133 Missense Mutation CGC,TGC R681C NP_066564.2
XM_006721817.3 2133 Missense Mutation CGC,TGC R682C XP_006721880.1
XM_006721818.3 2133 Missense Mutation CGC,TGC R321C XP_006721881.1
XM_017024448.1 2133 Missense Mutation CGC,TGC R320C XP_016879937.1

View Full Product Details