Product Details
- SNP ID
-
rs200260918
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:7626583 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGTTCCAGTCCAGGACGGCCAGG[A/C]GGAATTGGGAGCAGCCCTTATCCAA
- Phenotype
-
MIM: 611463
MIM: 182205
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SAT2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs13894] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SAT2
- Gene Name
- spermidine/spermine N1-acetyltransferase family member 2
- Gene
- SHBG
- Gene Name
- sex hormone binding globulin
View Full Product Details