Product Details

SNP ID
rs200329558
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4807797 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGACGGCGACCCCTGAGAGCCTC[C/T]TCCCCACTGGGGACGAACTGGACTC
Phenotype
MIM: 602384 MIM: 600307
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC107984973 PubMed Links

Gene Details

Gene
LOC107984973
Gene Name
atherin-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017025490.1 100 Intron XP_016880979.1
Gene
PLD2
Gene Name
phospholipase D2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243108.1 100 Missense Mutation CTC,TTC L9F NP_001230037.1
NM_002663.4 100 Missense Mutation CTC,TTC L9F NP_002654.3
XM_005256696.2 100 Intron XP_005256753.1
XM_017024764.1 100 UTR 5 XP_016880253.1
Gene
PSMB6
Gene Name
proteasome subunit beta 6
There are no transcripts associated with this gene.

View Full Product Details