Product Details

SNP ID
rs200398618
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48597827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCGAACACGCTCTTGTCCTGCGC[A/G]CAGTCCGACTTCCGCGGCTCGGGGT
Phenotype
MIM: 142966 MIM: 142960 MIM: 142961 MIM: 142962
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HOXB-AS3 PubMed Links

Gene Details

Gene
HOXB-AS3
Gene Name
HOXB cluster antisense RNA 3
There are no transcripts associated with this gene.

Gene
HOXB3
Gene Name
homeobox B3
There are no transcripts associated with this gene.

Gene
HOXB5
Gene Name
homeobox B5
There are no transcripts associated with this gene.

Gene
HOXB6
Gene Name
homeobox B6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018952.4 891 Silent Mutation TGC,TGT C108C NP_061825.2
XM_005257283.3 891 Silent Mutation TGC,TGT C108C XP_005257340.1
XM_005257284.3 891 Silent Mutation TGC,TGT C108C XP_005257341.1
XM_011524727.2 891 Silent Mutation TGC,TGT C172C XP_011523029.1
Gene
HOXB7
Gene Name
homeobox B7
There are no transcripts associated with this gene.

View Full Product Details