Product Details

SNP ID
rs200504646
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:47847378 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCTCCACTGCGCCGCCGGCCTTG[C/G]CCTCTCCCGAGGCCGCCCCAGCCGC
Phenotype
MIM: 614966 MIM: 608613
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LRRC46 PubMed Links

Gene Details

Gene
LRRC46
Gene Name
leucine rich repeat containing 46
There are no transcripts associated with this gene.

Gene
SCRN2
Gene Name
secernin 2
There are no transcripts associated with this gene.

Gene
SP6
Gene Name
Sp6 transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258248.1 1293 Missense Mutation GCC,GGC A351G NP_001245177.1
NM_199262.2 1293 Missense Mutation GCC,GGC A351G NP_954871.1
XM_006722115.3 1293 Missense Mutation GCC,GGC A351G XP_006722178.1

View Full Product Details