Product Details

SNP ID
rs199894904
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:63833418 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTGATGGACTCAATGGTCTCATG[C/G]ATCTGGAGAGGGTACCTGTGTCAGA
Phenotype
MIM: 601681 MIM: 601736
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FTSJ3 PubMed Links

Gene Details

Gene
FTSJ3
Gene Name
FtsJ homolog 3
There are no transcripts associated with this gene.

Gene
PSMC5
Gene Name
proteasome 26S subunit, ATPase 5
There are no transcripts associated with this gene.

Gene
SMARCD2
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098426.1 1184 Missense Mutation ATC,ATG I440M NP_001091896.1
XM_005257604.2 1184 Missense Mutation ATC,ATG I365M XP_005257661.2
XM_017024967.1 1184 Missense Mutation ATC,ATG I392M XP_016880456.1

View Full Product Details