Product Details

SNP ID
rs199520780
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:64858883 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATATCCTGGAAGTTCTTTTGTGA[A/G]CTAAAAAAAAAAAAACCAGAATGAG
Phenotype
MIM: 616557
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC37A3 PubMed Links

Gene Details

Gene
LRRC37A3
Gene Name
leucine rich repeat containing 37 member A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303255.1 5371 Missense Mutation NP_001290184.1
NM_199340.3 5371 Missense Mutation NP_955372.2
XM_011524776.1 5371 Intron XP_011523078.1
XM_011524777.1 5371 Intron XP_011523079.1
XM_011524778.2 5371 Intron XP_011523080.2
XM_017024593.1 5371 Intron XP_016880082.1
XM_017024594.1 5371 Intron XP_016880083.1
XM_017024595.1 5371 Intron XP_016880084.1
XM_017024596.1 5371 Intron XP_016880085.1
XM_017024597.1 5371 Intron XP_016880086.1
XM_017024598.1 5371 Intron XP_016880087.1
XM_017024599.1 5371 Intron XP_016880088.1
XM_017024600.1 5371 Intron XP_016880089.1
XM_017024601.1 5371 Missense Mutation CTC,TTC L833F XP_016880090.1
XM_017024602.1 5371 Missense Mutation CTC,TTC L758F XP_016880091.1
XM_017024603.1 5371 Intron XP_016880092.1
XM_017024604.1 5371 Intron XP_016880093.1
XM_017024605.1 5371 Intron XP_016880094.1
XM_017024606.1 5371 Intron XP_016880095.1
XM_017024607.1 5371 Intron XP_016880096.1
XM_017024608.1 5371 Intron XP_016880097.1

View Full Product Details