Product Details
- SNP ID
-
rs199966875
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:5183570 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTTCCCCCTACTGTGGATTCTCTG[A/G]TGCAGGACAAGGTTTGAGCTTCCCT
- Phenotype
-
MIM: 604334
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC100130950
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3853648] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC100130950
- Gene Name
- uncharacterized LOC100130950
There are no transcripts associated with this gene.
- Gene
- USP6
- Gene Name
- ubiquitin specific peptidase 6
There are no transcripts associated with this gene.
- Gene
- ZNF594
- Gene Name
- zinc finger protein 594
View Full Product Details