Product Details

SNP ID
rs200547902
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39160705 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCATGGAGTCCTTCACGTCCTGC[C/T]TATTGGCAAATATCAGGACTGAAGC
Phenotype
MIM: 606826
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARL5C PubMed Links

Gene Details

Gene
ARL5C
Gene Name
ADP ribosylation factor like GTPase 5C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143968.1 778 Missense Mutation AAG,AGG K126R NP_001137440.1
Gene
PLXDC1
Gene Name
plexin domain containing 1
There are no transcripts associated with this gene.

View Full Product Details