Product Details

SNP ID
rs200784238
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81683089 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCCAAGCTCCCGGATGGTGATC[C/T]TTCTCTGTGCCACGATGTCAGTAAG
Phenotype
MIM: 614271 MIM: 604375
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARL16 PubMed Links

Gene Details

Gene
ARL16
Gene Name
ADP ribosylation factor like GTPase 16
There are no transcripts associated with this gene.

Gene
CCDC137
Gene Name
coiled-coil domain containing 137
There are no transcripts associated with this gene.

Gene
HGS
Gene Name
hepatocyte growth factor-regulated tyrosine kinase substrate
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004712.4 Intron NP_004703.1
XM_011525463.2 Intron XP_011523765.1
XM_017025297.1 Intron XP_016880786.1
XM_017025298.1 Intron XP_016880787.1

View Full Product Details