Product Details

SNP ID
rs200817261
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:79834491 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGTGGTGATGGGGGTGCGGGTGC[A/G]GGTGGTGTGAGGGCGGGTGCGTGTT
Phenotype
MIM: 603079
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CBX4 PubMed Links

Gene Details

Gene
CBX4
Gene Name
chromobox 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003655.2 2823 Missense Mutation CCG,CTG P384L NP_003646.2
XM_011525399.2 2823 Missense Mutation CCG,CTG P318L XP_011523701.1

View Full Product Details