Product Details

SNP ID
rs201333261
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42762827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCATCACTCTTGTAGTATGGAGGA[A/G]TAAAGACAGTGAGGCCCAGGCCTAG
Phenotype
MIM: 605154
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAMP2 PubMed Links

Gene Details

Gene
RAMP2
Gene Name
receptor activity modifying protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005854.2 571 Missense Mutation AAT,AGT N168S NP_005845.2
Gene
RAMP2-AS1
Gene Name
RAMP2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details