Product Details

SNP ID
rs201607912
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28758379 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCAGCATGGCCAGGCTCTGTTCT[C/T]GGAAGCAGGCCTGTTGGAAATCCCC
Phenotype
MIM: 602464
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM222B PubMed Links

Gene Details

Gene
FAM222B
Gene Name
family with sequence similarity 222 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077498.2 1949 Missense Mutation NP_001070966.1
NM_001288631.1 1949 Missense Mutation NP_001275560.1
NM_001288632.1 1949 Missense Mutation NP_001275561.1
NM_001288633.1 1949 Missense Mutation NP_001275562.1
NM_001288634.1 1949 Missense Mutation NP_001275563.1
NM_001288635.1 1949 Missense Mutation NP_001275564.1
NM_001288636.1 1949 Missense Mutation NP_001275565.1
NM_001288637.1 1949 Missense Mutation NP_001275566.1
NM_001288638.1 1949 Missense Mutation NP_001275567.1
NM_001288639.1 1949 Missense Mutation NP_001275568.1
NM_001288640.1 1949 Missense Mutation NP_001275569.1
NM_018182.3 1949 Missense Mutation NP_060652.2
XM_011524986.2 1949 Missense Mutation XP_011523288.1
XM_011524987.1 1949 Missense Mutation XP_011523289.1
XM_011524988.2 1949 Missense Mutation XP_011523290.1
XM_017024831.1 1949 Missense Mutation XP_016880320.1
XM_017024832.1 1949 Missense Mutation XP_016880321.1
XM_017024833.1 1949 Missense Mutation XP_016880322.1
XM_017024834.1 1949 Missense Mutation XP_016880323.1
XM_017024835.1 1949 Missense Mutation XP_016880324.1
Gene
TRAF4
Gene Name
TNF receptor associated factor 4
There are no transcripts associated with this gene.

View Full Product Details