Product Details

SNP ID
rs201810212
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:78971595 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCGTCTAGTCCACACCTGAGGAG[C/T]TGGTCAGGTAGAAGGGGCGGATGAC
Phenotype
MIM: 600626
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LGALS3BP PubMed Links

Gene Details

Gene
LGALS3BP
Gene Name
galectin 3 binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005567.3 1918 Missense Mutation NP_005558.1

View Full Product Details