Product Details
- SNP ID
-
rs200890764
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:45137085 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTTGGTTACTTCGAGCCCCTGTAC[C/T]AGGTGATCCCTGACATGCCGAGGCC
- Phenotype
-
MIM: 608795
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ACBD4
PubMed Links
Gene Details
- Gene
- ACBD4
- Gene Name
- acyl-CoA binding domain containing 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001135705.2 |
1200 |
Nonsense Mutation |
CAG,TAG |
Q121* |
NP_001129177.1 |
NM_001135706.2 |
1200 |
Nonsense Mutation |
CAG,TAG |
Q121* |
NP_001129178.1 |
NM_001135707.2 |
1200 |
Nonsense Mutation |
CAG,TAG |
Q121* |
NP_001129179.1 |
NM_001321352.1 |
1200 |
Nonsense Mutation |
CAG,TAG |
Q121* |
NP_001308281.1 |
NM_001321353.1 |
1200 |
Nonsense Mutation |
CAG,TAG |
Q121* |
NP_001308282.1 |
NM_024722.3 |
1200 |
Nonsense Mutation |
CAG,TAG |
Q121* |
NP_078998.1 |
XM_006722085.2 |
1200 |
Nonsense Mutation |
CAG,TAG |
Q121* |
XP_006722148.1 |
XM_011525259.2 |
1200 |
Nonsense Mutation |
CAG,TAG |
Q121* |
XP_011523561.1 |
XM_011525261.2 |
1200 |
Nonsense Mutation |
CAG,TAG |
Q121* |
XP_011523563.1 |
XM_017025084.1 |
1200 |
Nonsense Mutation |
CAG,TAG |
Q121* |
XP_016880573.1 |
XM_017025085.1 |
1200 |
Nonsense Mutation |
CAG,TAG |
Q121* |
XP_016880574.1 |
XM_017025086.1 |
1200 |
Nonsense Mutation |
CAG,TAG |
Q121* |
XP_016880575.1 |
XM_017025087.1 |
1200 |
Nonsense Mutation |
CAG,TAG |
Q121* |
XP_016880576.1 |
XM_017025088.1 |
1200 |
Nonsense Mutation |
CAG,TAG |
Q121* |
XP_016880577.1 |
XM_017025089.1 |
1200 |
Nonsense Mutation |
CAG,TAG |
Q121* |
XP_016880578.1 |
XM_017025090.1 |
1200 |
Nonsense Mutation |
CAG,TAG |
Q121* |
XP_016880579.1 |
XM_017025091.1 |
1200 |
Nonsense Mutation |
CAG,TAG |
Q121* |
XP_016880580.1 |
XM_017025092.1 |
1200 |
Nonsense Mutation |
CAG,TAG |
Q121* |
XP_016880581.1 |
XM_017025093.1 |
1200 |
Nonsense Mutation |
CAG,TAG |
Q121* |
XP_016880582.1 |
XM_017025094.1 |
1200 |
Nonsense Mutation |
CAG,TAG |
Q121* |
XP_016880583.1 |
XM_017025095.1 |
1200 |
Nonsense Mutation |
CAG,TAG |
Q121* |
XP_016880584.1 |
XM_017025096.1 |
1200 |
Nonsense Mutation |
CAG,TAG |
Q121* |
XP_016880585.1 |
XM_017025097.1 |
1200 |
Nonsense Mutation |
CAG,TAG |
Q121* |
XP_016880586.1 |
XM_017025098.1 |
1200 |
Nonsense Mutation |
CAG,TAG |
Q121* |
XP_016880587.1 |
XM_017025099.1 |
1200 |
Nonsense Mutation |
CAG,TAG |
Q121* |
XP_016880588.1 |
XM_017025100.1 |
1200 |
Nonsense Mutation |
CAG,TAG |
Q121* |
XP_016880589.1 |
XM_017025101.1 |
1200 |
Nonsense Mutation |
CAG,TAG |
Q121* |
XP_016880590.1 |
- Gene
- PLCD3
- Gene Name
- phospholipase C delta 3
There are no transcripts associated with this gene.
View Full Product Details