Product Details
- SNP ID
-
rs200685037
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:41190184 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGGTCCAGCTGCTGTGGCCAAACC[A/T]GCTGTGGGTCCAGCTGCTGTCAGCC
- Phenotype
-
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
KRTAP4-1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs238824] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KRTAP4-1
- Gene Name
- keratin associated protein 4-1
There are no transcripts associated with this gene.
- Gene
- KRTAP9-1
- Gene Name
- keratin associated protein 9-1
View Full Product Details