Product Details

SNP ID
rs201132553
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1725642 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGCTGAGGCCTTGCTGGAGTCGC[C/T]GGAGATGCTGTATGTGGTACACCCT
Phenotype
MIM: 614218
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR22HG PubMed Links

Gene Details

Gene
MIR22HG
Gene Name
MIR22 host gene
There are no transcripts associated with this gene.

Gene
WDR81
Gene Name
WD repeat domain 81
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163673.1 812 Intron NP_001157145.1
NM_001163809.1 812 Missense Mutation CCG,CTG P228L NP_001157281.1
NM_001163811.1 812 Intron NP_001157283.1
NM_152348.3 812 Intron NP_689561.2
XM_011523651.2 812 Intron XP_011521953.1
XM_017024184.1 812 Missense Mutation CCG,CTG P228L XP_016879673.1

View Full Product Details