Product Details

SNP ID
rs201506936
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8289305 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGTGCAGGGGGCTAGGGCTGTCC[A/G]TGTGGAGTCTGTTCAGCCTCTCAGT
Phenotype
MIM: 607954 MIM: 610818
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RANGRF PubMed Links

Gene Details

Gene
RANGRF
Gene Name
RAN guanine nucleotide release factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177801.1 380 Missense Mutation CAT,CGT H81R NP_001171272.1
NM_001177802.1 380 Missense Mutation CAT,CGT H81R NP_001171273.1
NM_016492.4 380 Missense Mutation CAT,CGT H81R NP_057576.2
XM_005256618.4 380 Missense Mutation CAT,CGT H81R XP_005256675.1
Gene
SLC25A35
Gene Name
solute carrier family 25 member 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320870.1 380 Intron NP_001307799.1
NM_001320871.1 380 Intron NP_001307800.1
NM_001320872.1 380 UTR 3 NP_001307801.1
NM_001320877.1 380 Intron NP_001307806.1
NM_201520.2 380 UTR 3 NP_958928.1
XM_005256641.2 380 Intron XP_005256698.1
XM_011523846.1 380 Intron XP_011522148.1
XM_017024633.1 380 Intron XP_016880122.1
XM_017024634.1 380 Intron XP_016880123.1

View Full Product Details