Product Details

SNP ID
rs201646494
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81686345 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCTGTGAATTCCATCAAGAAGAA[A/G]GTCAACGACAAGAACCCACACGTCG
Phenotype
MIM: 604375
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARL16 PubMed Links

Gene Details

Gene
ARL16
Gene Name
ADP ribosylation factor like GTPase 16
There are no transcripts associated with this gene.

Gene
HGS
Gene Name
hepatocyte growth factor-regulated tyrosine kinase substrate
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004712.4 240 Silent Mutation AAA,AAG K52K NP_004703.1
XM_011525463.2 240 Silent Mutation AAA,AAG K52K XP_011523765.1
XM_017025297.1 240 Silent Mutation AAA,AAG K43K XP_016880786.1
XM_017025298.1 240 UTR 5 XP_016880787.1
Gene
MIR6786
Gene Name
microRNA 6786
There are no transcripts associated with this gene.

View Full Product Details