Product Details
- SNP ID
-
rs200823933
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:75627692 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGCGGGCAAAGCCTTTAAACAACT[C/T]CTGGAAGGGAGAGGGAGAAGAGAAG
- Phenotype
-
MIM: 603781
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MYO15B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs376539887] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MYO15B
- Gene Name
- myosin XVB
- Gene
- RECQL5
- Gene Name
- RecQ like helicase 5
- Gene
- SMIM5
- Gene Name
- small integral membrane protein 5
There are no transcripts associated with this gene.
View Full Product Details