Product Details

SNP ID
rs201136101
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3716741 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACACCAGAAGTCCACCAACGCTGC[A/C]TTTAATGATGATAGGCAAAGAATGG
Phenotype
MIM: 609240 MIM: 604682
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GSG2 PubMed Links

Gene Details

Gene
GSG2
Gene Name
germ cell associated 2, haspin
There are no transcripts associated with this gene.

Gene
ITGAE
Gene Name
integrin subunit alpha E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002208.4 3821 Missense Mutation GGC,TGC G1131C NP_002199.3
XM_011523823.2 3821 Missense Mutation GGC,TGC G1169C XP_011522125.1
XM_011523825.2 3821 Missense Mutation GGC,TGC G1137C XP_011522127.1
XM_011523827.2 3821 Missense Mutation GGC,TGC G1115C XP_011522129.1
XM_011523828.2 3821 Missense Mutation GGC,TGC G1089C XP_011522130.1
XM_017024586.1 3821 Missense Mutation GGC,TGC G1143C XP_016880075.1
XM_017024587.1 3821 Intron XP_016880076.1

View Full Product Details