Product Details

SNP ID
rs201854089
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:66878980 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTCTTGTCTCCACAGGTGAGGAG[C/T]GGGGGCGTTGCTTCACCATAGAATA
Phenotype
MIM: 606405
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CACNG5 PubMed Links

Gene Details

Gene
CACNG5
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145811.2 265 Missense Mutation CGG,TGG R69W NP_665810.1

View Full Product Details