Product Details

SNP ID
rs202165065
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7392834 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCACAGCCACAGGTCCAGCAGGGC[C/T]CCACCACTCGCAAGACTGTCTGGCG
Phenotype
MIM: 607611 MIM: 608076
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PLSCR3 PubMed Links

Gene Details

Gene
PLSCR3
Gene Name
phospholipid scramblase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201576.1 1150 Missense Mutation GAG,GGG E209G NP_001188505.1
NM_020360.3 1150 Missense Mutation GAG,GGG E209G NP_065093.2
Gene
TMEM256-PLSCR3
Gene Name
TMEM256-PLSCR3 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
TNK1
Gene Name
tyrosine kinase non receptor 1
There are no transcripts associated with this gene.

View Full Product Details