Product Details

SNP ID
rs200826404
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:5210928 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTATTTACCAGTGAGTATGTAGCG[G/T]GCGGCTGACTTGGGGCTTCCTGTGG
Phenotype
MIM: 614406
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC100130950 PubMed Links

Gene Details

Gene
LOC100130950
Gene Name
uncharacterized LOC100130950
There are no transcripts associated with this gene.

Gene
SCIMP
Gene Name
SLP adaptor and CSK interacting membrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271842.1 416 Missense Mutation CAC,CCC H97P NP_001258771.1
NM_001319190.1 416 Intron NP_001306119.1
NM_207103.3 416 Missense Mutation CAC,CCC H104P NP_996986.1
XM_017024615.1 416 Intron XP_016880104.1

View Full Product Details