Product Details
- SNP ID
-
rs201341928
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:18245170 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGGCCTTGCAGAAGGCGCTCCAGG[C/T]GTGGAAGCAGCGGGTAAAGGCGTGC
- Phenotype
-
MIM: 600362
MIM: 600966
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FLII
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7498] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FLII
- Gene Name
- FLII, actin remodeling protein
- Gene
- LLGL1
- Gene Name
- LLGL1, scribble cell polarity complex component
View Full Product Details