Product Details
- SNP ID
-
rs201375413
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:63940858 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGCAGTGTCTGAGGGGCTGATGGG[C/T]ATCAGCCCCATAGTGGGTCCGGCGT
- Phenotype
-
MIM: 147245
MIM: 603967
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CD79B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2227907] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CD79B
- Gene Name
- CD79b molecule
There are no transcripts associated with this gene.
- Gene
- SCN4A
- Gene Name
- sodium voltage-gated channel alpha subunit 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000334.4 |
5501 |
Missense Mutation |
ATA,ATG |
I1808M |
NP_000325.4 |
View Full Product Details