Product Details

SNP ID
rs201715422
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:50278599 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAATGCTGTGGTGACAGCTGCTGC[C/G]AGGAGAACGAGCTCTTCCCTGGCCC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TMEM92 PubMed Links

Gene Details

Gene
TMEM92
Gene Name
transmembrane protein 92
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168215.1 249 Missense Mutation CAG,GAG Q47E NP_001161687.1
NM_153229.2 249 Missense Mutation CAG,GAG Q47E NP_694961.2
XM_006721730.3 249 Missense Mutation CAG,GAG Q47E XP_006721793.1
Gene
TMEM92-AS1
Gene Name
TMEM92 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details