Product Details

SNP ID
rs201716457
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:6652026 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGTCCCACCGCGTGGAGACGTCGT[C/G]CCGGCGGGTGGAGACCTCCCAGCGC
Phenotype
MIM: 616967
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ALOX15P1 PubMed Links

Gene Details

Gene
ALOX15P1
Gene Name
arachidonate 15-lipoxygenase pseudogene 1
There are no transcripts associated with this gene.

Gene
C17orf100
Gene Name
chromosome 17 open reading frame 100
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105520.1 288 Missense Mutation TCC,TGC S38C NP_001098990.1
Gene
MED31
Gene Name
mediator complex subunit 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016060.2 288 Intron NP_057144.1
XM_017024710.1 288 Intron XP_016880199.1
Gene
MIR4520-1
Gene Name
microRNA 4520-1
There are no transcripts associated with this gene.

Gene
MIR4520-2
Gene Name
microRNA 4520-2
There are no transcripts associated with this gene.

Gene
TXNDC17
Gene Name
thioredoxin domain containing 17
There are no transcripts associated with this gene.

View Full Product Details