Product Details
- SNP ID
-
rs201785449
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:40661968 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGCAGGTGGTGCCACTGGCGTCGG[A/G]CCTCCTTGAGTTCTGCTTGAGCTGC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRT222
PubMed Links
Gene Details
- Gene
- KRT222
- Gene Name
- keratin 222
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152349.2 |
238 |
Missense Mutation |
GCC,GTC |
A58V |
NP_689562.1 |
- Gene
- LOC105371775
- Gene Name
- uncharacterized LOC105371775
There are no transcripts associated with this gene.
View Full Product Details