Product Details

SNP ID
rs200724643
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:27882590 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACCTGCCTGACAGCATGCTTGTA[A/G]TGCTGCTGGATGCCCTTGGTCGGCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LYRM9 PubMed Links

Gene Details

Gene
LYRM9
Gene Name
LYR motif containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001076680.1 190 Silent Mutation CAC,CAT H35H NP_001070148.1
XM_011524491.2 190 Silent Mutation CAC,CAT H61H XP_011522793.1
XM_017024325.1 190 Silent Mutation CAC,CAT H35H XP_016879814.1

View Full Product Details