Product Details

SNP ID
rs200725590
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:45024697 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGGTTCAGGCTGTTCCGCCGCAT[C/T]AGCCGTGCCAGCTGTGTGTCCCGGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCAKD PubMed Links

Gene Details

Gene
DCAKD
Gene Name
dephospho-CoA kinase domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128631.2 788 Silent Mutation CTA,CTG L144L NP_001122103.1
NM_001288654.2 788 Silent Mutation CTA,CTG L144L NP_001275583.1
NM_001288655.1 788 Silent Mutation CTA,CTG L144L NP_001275584.1
NM_001321326.1 788 Silent Mutation CTA,CTG L144L NP_001308255.1
NM_024819.6 788 Silent Mutation CTA,CTG L144L NP_079095.3
XM_005257688.2 788 Intron XP_005257745.1
XM_011525262.2 788 Silent Mutation CTA,CTG L144L XP_011523564.1
XM_017025102.1 788 Intron XP_016880591.1
XM_017025103.1 788 Intron XP_016880592.1
XM_017025104.1 788 Intron XP_016880593.1
XM_017025105.1 788 Silent Mutation CTA,CTG L43L XP_016880594.1
XM_017025106.1 788 Silent Mutation CTA,CTG L43L XP_016880595.1

View Full Product Details