Product Details

SNP ID
rs201924207
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:50353871 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGCCGCCAGAACCTGAGTGGGG[A/C]AGCAGCTGGGGAGGCGCTGGTAGGG
Phenotype
MIM: 608125
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
XYLT2 PubMed Links

Gene Details

Gene
XYLT2
Gene Name
xylosyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022167.3 800 Missense Mutation GAA,GCA E126A NP_071450.2
XM_005257572.4 800 Missense Mutation GAA,GCA E94A XP_005257629.1

View Full Product Details