Product Details

SNP ID
rs202135672
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74473564 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTTGCTCTGAGCAAATGCAGGAC[C/T]GTGGCGGGCCCCGTGGGGGGATCCC
Phenotype
MIM: 606790
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD300A PubMed Links

Gene Details

Gene
CD300A
Gene Name
CD300a molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256841.1 413 Intron NP_001243770.1
NM_007261.3 413 Silent Mutation ACC,ACT T23T NP_009192.2
XM_005256990.2 413 Silent Mutation ACC,ACT T23T XP_005257047.1
XM_005256991.4 413 Silent Mutation ACC,ACT T23T XP_005257048.1
XM_005256992.4 413 UTR 5 XP_005257049.1
XM_006721656.3 413 UTR 5 XP_006721719.1
XM_011524254.2 413 Intron XP_011522556.1

View Full Product Details