Product Details

SNP ID
rs200660734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76007369 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGATGCGGCCTGTCCTCTTGGGGT[C/T]GATGAGCCCCCCGGTCAGGTGCTGC
Phenotype
MIM: 123828 MIM: 601590 MIM: 613130
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDK3 PubMed Links

Gene Details

Gene
CDK3
Gene Name
cyclin dependent kinase 3
There are no transcripts associated with this gene.

Gene
EVPL
Gene Name
envoplakin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320747.1 6156 Missense Mutation AAC,GAC N1968D NP_001307676.1
NM_001988.3 6156 Missense Mutation AAC,GAC N1946D NP_001979.2
Gene
TEN1
Gene Name
TEN1 CST complex subunit
There are no transcripts associated with this gene.

Gene
TEN1-CDK3
Gene Name
TEN1-CDK3 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details