Product Details
- SNP ID
-
rs200829899
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:73169505 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCATCATTGGGTTGTGTGGCAACAC[A/T]CTTGTCATTTATGTCATCCTCCGCT
- Phenotype
-
MIM: 182452
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SSTR2
PubMed Links
Gene Details
- Gene
- SSTR2
- Gene Name
- somatostatin receptor 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001050.2 |
546 |
Silent Mutation |
ACA,ACT |
T62T |
NP_001041.1 |
View Full Product Details