Product Details

SNP ID
rs202139169
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74582290 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCTTGCTGTTGTGGTTGTCCAT[C/T]CTGAGACTGCAGTTTGTGTGCCTAA
Phenotype
MIM: 616301
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
C17orf77 PubMed Links

Gene Details

Gene
C17orf77
Gene Name
chromosome 17 open reading frame 77
There are no transcripts associated with this gene.

Gene
CD300LD
Gene Name
CD300 molecule like family member d
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001115152.1 430 Missense Mutation GAA,GGA E134G NP_001108624.1

View Full Product Details