Product Details

SNP ID
rs200799100
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:18245162 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTAGGCCAGGGCCTTGCAGAAGGC[A/G]CTCCAGGCGTGGAAGCAGCGGGTAA
Phenotype
MIM: 600362 MIM: 600966
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FLII PubMed Links

Gene Details

Gene
FLII
Gene Name
FLII, actin remodeling protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256264.1 3798 Silent Mutation AGC,AGT S1251S NP_001243193.1
NM_001256265.1 3798 Silent Mutation AGC,AGT S1207S NP_001243194.1
NM_002018.3 3798 Silent Mutation AGC,AGT S1262S NP_002009.1
XM_005256555.3 3798 Silent Mutation AGC,AGT S1289S XP_005256612.1
XM_005256556.3 3798 Silent Mutation AGC,AGT S1288S XP_005256613.1
XM_005256558.2 3798 Silent Mutation AGC,AGT S1261S XP_005256615.1
Gene
LLGL1
Gene Name
LLGL1, scribble cell polarity complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004140.3 3798 Intron NP_004131.3
XM_011523849.2 3798 Intron XP_011522151.1
XM_011523850.2 3798 Intron XP_011522152.1
XM_011523851.2 3798 Intron XP_011522153.1
XM_011523852.2 3798 Intron XP_011522154.1
XM_011523853.2 3798 Intron XP_011522155.1
XM_011523854.2 3798 Intron XP_011522156.1
XM_011523855.2 3798 Intron XP_011522157.1
XM_011523856.1 3798 Intron XP_011522158.1

View Full Product Details